Search
Search

E2F6| Gene Symbol: E2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266655.1 | NP_001253584.1 | 366 | 99 | ||
| XM_015111694.2 | XP_014967180.1 | 401 | 99 | |||
| GenBank mRNA | JU474575.1 | - | 255 | 99 | ||
| JV046365.1 | - | 402 | 99 | |||
| JV635866.1 | - | 366 | 99 | |||
Set Membership: |
|