Search
Search

NEBL| Gene Symbol: NEBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146577.2 | XP_015002063.2 | 3371 | 80 | ||
| XM_015146581.2 | XP_015002067.2 | 3128 | 80 | |||
| XM_015146582.2 | XP_015002068.1 | 983 | 80 | |||
| XM_028826044.1 | XP_028681877.1 | 2896 | 80 | |||
| XM_028826045.1 | XP_028681878.1 | 3266 | 80 | |||
| GenBank mRNA | JU320560.1 | - | 922 | 80 | ||
| JV703620.1 | - | 864 | 80 | |||
Set Membership: |
|