Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02892979_m1
See other DEPDC5 GE Assays ›
Gene Symbol:
DEPDC5
Gene Name:
DEP domain containing 5, GATOR1 subcomplex subunit
Gene Aliases:
EGK_02963
Chromosome Location:
Chr.10: 19594025 - 19758791 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02892979_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DEPDC5
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015150093.2
XP_015005579.2
765
109
XM_015150094.2
XP_015005580.2
669
109
XM_015150096.2
XP_015005582.2
765
109
XM_015150099.2
XP_015005585.2
765
109
XM_015150100.2
XP_015005586.2
767
109
XM_015150101.2
XP_015005587.2
669
109
XM_028827770.1
XP_028683603.1
767
109
XM_028827771.1
XP_028683604.1
669
109
XM_028827772.1
XP_028683605.1
767
109
XM_028827773.1
XP_028683606.1
767
109
XM_028827774.1
XP_028683607.1
767
109
XM_028827775.1
XP_028683608.1
669
109
XM_028827776.1
XP_028683609.1
767
109
XM_028827777.1
XP_028683610.1
669
109
XM_028827778.1
XP_028683611.1
767
109
XM_028827779.1
XP_028683612.1
669
109
XM_028827780.1
XP_028683613.1
767
109
XM_028827781.1
XP_028683614.1
669
109
XM_028827782.1
XP_028683615.1
767
109
XM_028827783.1
XP_028683616.1
767
109
XM_028827784.1
XP_028683617.1
767
109
GenBank mRNA
JU337272.1
-
623
109
JV048812.1
-
705
109
JV637100.1
-
691
109
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Macaca mulatta
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline