Search
Search

COL11A1| Gene Symbol: COL11A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146004.2 | XP_015001490.1 | 2777 | 109 | ||
| XM_015146012.2 | XP_015001498.1 | 2771 | 109 | |||
| XM_015146016.2 | XP_015001502.1 | 2624 | 109 | |||
| GenBank mRNA | JU332934.1 | - | 2422 | 109 | ||
Set Membership: |
|