Search
Search

IK| Gene Symbol: IK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256897.2 | NP_001243826.1 | 1468 | 80 | ||
| GenBank mRNA | JU320589.1 | - | 1452 | 80 | ||
| JU472994.1 | - | 1452 | 80 | |||
| JV490075.1 | - | 1471 | 80 | |||
Set Membership: |
|