Search
Search

LAMTOR4| Gene Symbol: LAMTOR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134433.2 | XP_014989919.1 | 336 | 114 | ||
| XM_028845572.1 | XP_028701405.1 | 223 | 114 | |||
| GenBank mRNA | JU334460.1 | - | 270 | 114 | ||
| JU475549.1 | - | 262 | 114 | |||
| JV045794.1 | - | 270 | 114 | |||
Set Membership: |
|