Search
Search

PNOC| Gene Symbol: PNOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261798.1 | NP_001248727.1 | 125 | 122 | ||
| XM_015145032.2 | XP_015000518.2 | 419 | 122 | |||
| XM_015145033.2 | XP_015000519.2 | 428 | 122 | |||
| XM_015145034.2 | XP_015000520.2 | 419 | 122 | |||
| GenBank mRNA | JU335036.1 | - | 125 | 122 | ||
| JV548141.1 | - | 201 | 122 | |||
Set Membership: |
|