Search
Search

SLC35B1| Gene Symbol: SLC35B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119333.2 | XP_014974819.1 | 540 | 64 | ||
| GenBank mRNA | JU319813.1 | - | 300 | 64 | ||
| JU473041.1 | - | 279 | 64 | |||
| JV044292.1 | - | 291 | 64 | |||
Set Membership: |
|