Search
Search

TVP23B| Gene Symbol: TVP23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260706.1 | NP_001247635.1 | 390 | 107 | ||
| XM_015118616.2 | XP_014974102.1 | 631 | 107 | |||
| XM_015118617.2 | XP_014974103.1 | 520 | 107 | |||
| XM_015118618.2 | XP_014974104.1 | 522 | 107 | |||
| XM_015118619.2 | XP_014974105.1 | 558 | 107 | |||
| XM_015118620.2 | XP_014974106.1 | 583 | 107 | |||
| XM_015118621.2 | XP_014974107.1 | 518 | 107 | |||
| XM_015118622.2 | XP_014974108.1 | 278 | 107 | |||
| XM_028835463.1 | XP_028691296.1 | 1541 | 107 | |||
| XM_028835464.1 | XP_028691297.1 | 631 | 107 | |||
| GenBank mRNA | AY680486.1 | - | 228 | 107 | ||
| JU322751.1 | - | 435 | 107 | |||
| JU475807.1 | - | 353 | 107 | |||
| JV047232.1 | - | 390 | 107 | |||
Set Membership: |
|