Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02921881_g1
See other SEC31B GE Assays ›
Gene Symbol:
SEC31B
Gene Name:
SEC31 homolog B, COPII coat complex component
Gene Aliases:
-
Chromosome Location:
Chr.9: 100950475 - 100987257 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02921881_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SEC31B
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015147925.2
XP_015003411.2
2082
82
XM_015147926.2
XP_015003412.2
2025
82
XM_015147927.2
XP_015003413.2
2319
82
XM_015147928.2
XP_015003414.2
1980
82
XM_015147929.2
XP_015003415.2
2282
82
XM_015147930.2
XP_015003416.2
2079
82
XM_015147931.2
XP_015003417.2
2082
82
XM_015147932.2
XP_015003418.2
2073
82
XM_015147933.2
XP_015003419.2
2073
82
XM_015147934.2
XP_015003420.2
1970
82
XM_015147935.2
XP_015003421.2
2082
82
XM_015147936.2
XP_015003422.2
2070
82
XM_015147937.2
XP_015003423.2
2064
82
XM_015147938.2
XP_015003424.2
2082
82
XM_015147939.2
XP_015003425.2
2061
82
XM_015147940.2
XP_015003426.2
1958
82
XM_015147941.2
XP_015003427.2
2082
82
XM_015147945.2
XP_015003431.2
2082
82
XM_015147946.2
XP_015003432.2
1856
82
XM_015147947.2
XP_015003433.2
2158
82
XM_015147948.2
XP_015003434.2
1791
82
XM_015147949.2
XP_015003435.2
2082
82
XM_015147951.2
XP_015003437.2
1820
82
XM_015147952.2
XP_015003438.2
1790
82
XM_028826757.1
XP_028682590.1
2070
82
XM_028826758.1
XP_028682591.1
1854
82
XM_028826759.1
XP_028682592.1
1894
82
XM_028826760.1
XP_028682593.1
2082
82
XM_028826761.1
XP_028682594.1
1880
82
XM_028826762.1
XP_028682595.1
1901
82
XM_028826763.1
XP_028682596.1
2195
82
XM_028826764.1
XP_028682597.1
2082
82
XM_028826765.1
XP_028682598.1
2082
82
XM_028826766.1
XP_028682599.1
2082
82
XM_028826767.1
XP_028682600.1
2082
82
XM_028826768.1
XP_028682601.1
2082
82
XM_028826769.1
XP_028682602.1
1588
82
GenBank mRNA
JV647750.1
-
1088
82
JV711551.1
-
1070
82
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Recommended Assay
Macaca mulatta
Panther Classification:
Molecular Function -
vesicle coat protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline