Search
Search

ANAPC5| Gene Symbol: ANAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152986.1 | XP_015008472.1 | 2169 | 80 | ||
| XM_015152987.2 | XP_015008473.1 | 2135 | 80 | |||
| XM_015152988.2 | XP_015008474.1 | 2294 | 80 | |||
| XM_015152989.2 | XP_015008475.1 | 2640 | 80 | |||
| XM_028830001.1 | XP_028685834.1 | 2046 | 80 | |||
| GenBank mRNA | JU334892.1 | - | 2072 | 80 | ||
| JU334893.1 | - | 2033 | 80 | |||
Set Membership: |
|