Search
Search

MANBAL| Gene Symbol: MANBAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266836.1 | NP_001253765.1 | 251 | 154 | ||
| XM_015149038.2 | XP_015004524.1 | 340 | 154 | |||
| GenBank mRNA | JU320124.1 | - | 244 | 154 | ||
| JU476742.1 | - | 245 | 154 | |||
| JV045752.1 | - | 251 | 154 | |||
Set Membership: |
|