Search
Search

PRPF3| Gene Symbol: PRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028829395.1 | XP_028685228.1 | 909 | 97 | ||
| XM_028829396.1 | XP_028685229.1 | 1403 | 97 | |||
| GenBank mRNA | JU337047.1 | - | 837 | 97 | ||
| JU477776.1 | - | 877 | 97 | |||
| JV043557.1 | - | 886 | 97 | |||
Set Membership: |
|