Search
Search

BOLA2B| Gene Symbol: BOLA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194470.3 | NP_001181399.2 | 545 | 150 | ||
| XM_015125897.2 | XP_014981383.2 | 621 | 150 | |||
| XM_028840539.1 | XP_028696372.1 | 179 | 150 | |||
Set Membership: |
|