Search
Search

SEC14L2| Gene Symbol: SEC14L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150145.2 | XP_015005631.1 | 1166 | 84 | ||
| XM_015150147.2 | XP_015005633.1 | 1166 | 84 | |||
| XM_015150150.2 | XP_015005636.1 | 1007 | 84 | |||
| XM_028827846.1 | XP_028683679.1 | 950 | 84 | |||
| GenBank mRNA | JU333723.1 | - | 1004 | 84 | ||
Set Membership: |
|