Search
Search

TLCD2| Gene Symbol: TLCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194730.2 | NP_001181659.2 | 340 | 83 | ||
| XM_028835774.1 | XP_028691607.1 | 5828 | 83 | |||
| XM_028835775.1 | XP_028691608.1 | 4425 | 83 | |||
| XM_028835776.1 | XP_028691609.1 | 4340 | 83 | |||
| XM_028835777.1 | XP_028691610.1 | 5048 | 83 | |||
| XM_028835778.1 | XP_028691611.1 | 4901 | 83 | |||
| XM_028835780.1 | XP_028691613.1 | 3506 | 83 | |||
| XM_028835781.1 | XP_028691614.1 | 3409 | 83 | |||
Set Membership: |
|