Search
Search

APLN| Gene Symbol: APLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190868.1 | NP_001177797.1 | 74 | 61 | ||
| XM_015128162.2 | XP_014983648.1 | 493 | 61 | |||
| GenBank mRNA | JU336931.1 | - | 347 | 61 | ||
Set Membership: |
|