Search
Search

BEST1| Gene Symbol: BEST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116583.4 | XP_001116583.2 | 1029 | 76 | ||
| XM_015113980.2 | XP_014969466.2 | 1187 | 76 | |||
| XM_015113983.2 | XP_014969469.2 | 1108 | 76 | |||
| GenBank mRNA | JU325185.1 | - | 159 | 76 | ||
Set Membership: |
|