Search
Search

MAPRE3| Gene Symbol: MAPRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111936.2 | XP_014967422.1 | 916 | 109 | ||
| XM_028831861.1 | XP_028687694.1 | 708 | 109 | |||
| GenBank mRNA | JU333484.1 | - | 842 | 109 | ||
| JV490902.1 | - | 633 | 109 | |||
| JV528210.1 | - | 622 | 109 | |||
| JV635549.1 | - | 513 | 109 | |||
Set Membership: |
|