Search
Search

OGFOD1| Gene Symbol: OGFOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841321.1 | XP_028697154.1 | 1511 | 103 | ||
| XM_028841322.1 | XP_028697155.1 | 1711 | 103 | |||
| GenBank mRNA | JU335281.1 | - | 1486 | 103 | ||
| JV718094.1 | - | 1474 | 103 | |||
Set Membership: |
|