Search
Search

SRPK3| Gene Symbol: SRPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128522.2 | XP_014984008.2 | 1619 | 65 | ||
| XM_028842311.1 | XP_028698144.1 | 1623 | 65 | |||
| GenBank mRNA | JU326254.1 | - | 1352 | 65 | ||
| JV578279.1 | - | 1397 | 65 | |||
Set Membership: |
|