Search
Search

CDH15| Gene Symbol: CDH15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126803.2 | XP_014982289.2 | 1459 | 97 | ||
| XM_015126804.2 | XP_014982290.2 | 1461 | 97 | |||
| GenBank mRNA | JV639744.1 | - | 116 | 97 | ||
Set Membership: |
|