Search
Search

ARHGEF7| Gene Symbol: ARHGEF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121452.2 | XP_014976938.1 | 2267 | 71 | ||
| XM_015121454.2 | XP_014976940.1 | 2265 | 71 | |||
| XM_015121456.2 | XP_014976942.1 | 1347 | 71 | |||
| XM_015121459.2 | XP_014976945.1 | 906 | 71 | |||
| XM_015121461.2 | XP_014976947.1 | 1009 | 71 | |||
| XM_015121462.2 | XP_014976948.1 | 793 | 71 | |||
| XM_028837324.1 | XP_028693157.1 | 1347 | 71 | |||
| XM_028837325.1 | XP_028693158.1 | 1347 | 71 | |||
| XM_028837326.1 | XP_028693159.1 | 1131 | 71 | |||
| XM_028837327.1 | XP_028693160.1 | 1347 | 71 | |||
| XM_028837328.1 | XP_028693161.1 | 909 | 71 | |||
| XM_028837329.1 | XP_028693162.1 | 1010 | 71 | |||
| XM_028837330.1 | XP_028693163.1 | 880 | 71 | |||
| XM_028837331.1 | XP_028693164.1 | 795 | 71 | |||
| XM_028837332.1 | XP_028693165.1 | 742 | 71 | |||
| XM_028837333.1 | XP_028693166.1 | 737 | 71 | |||
| XM_028837334.1 | XP_028693167.1 | 1131 | 71 | |||
| XM_028837335.1 | XP_028693168.1 | 2058 | 71 | |||
| XM_028837336.1 | XP_028693169.1 | 1008 | 71 | |||
| XM_028837337.1 | XP_028693170.1 | 500 | 71 | |||
| XM_028837338.1 | XP_028693171.1 | 794 | 71 | |||
| GenBank mRNA | JU336932.1 | - | 1164 | 71 | ||
| JU350058.1 | - | 1164 | 71 | |||
Set Membership: |
|