Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh06008377_m1
See other ZMYND8 GE Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
-
Chromosome Location:
Chr.10: 82404821 - 82553674 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh06008377_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015148870.2
XP_015004356.1
3129
63
XM_015148871.2
XP_015004357.1
3231
63
XM_015148872.2
XP_015004358.1
3014
63
XM_015148873.2
XP_015004359.1
3308
63
XM_015148874.2
XP_015004360.1
3216
63
XM_015148876.2
XP_015004362.1
3156
63
XM_015148877.2
XP_015004363.1
3231
63
XM_015148878.2
XP_015004364.1
3141
63
XM_015148879.2
XP_015004365.1
3105
63
XM_015148880.2
XP_015004366.1
3093
63
XM_015148881.2
XP_015004367.1
3308
63
XM_015148882.2
XP_015004368.1
3156
63
XM_015148883.2
XP_015004369.1
3170
63
XM_015148884.2
XP_015004370.1
3018
63
XM_015148885.2
XP_015004371.1
3093
63
XM_015148887.2
XP_015004373.1
2957
63
XM_015148888.2
XP_015004374.1
3018
63
XM_028828476.1
XP_028684309.1
3057
63
XM_028828477.1
XP_028684310.1
2978
63
XM_028828478.1
XP_028684311.1
3129
63
XM_028828479.1
XP_028684312.1
3780
63
XM_028828480.1
XP_028684313.1
2991
63
XM_028828481.1
XP_028684314.1
2978
63
XM_028828482.1
XP_028684315.1
2991
63
XM_028828484.1
XP_028684317.1
2840
63
GenBank mRNA
JU321123.1
-
2849
63
JU321146.1
-
2924
63
JU474984.1
-
2851
63
JU474985.1
-
2926
63
JV043154.1
-
2920
63
JV043155.1
-
2920
63
JV046619.1
-
2914
63
JV046620.1
-
2845
63
JV647087.1
-
2800
63
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Macaca mulatta
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit