Search
Search

TRIM66| Gene Symbol: TRIM66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114920.2 | XP_014970406.2 | 1572 | 76 | ||
| XM_015114921.2 | XP_014970407.2 | 1572 | 76 | |||
| XM_015114922.2 | XP_014970408.2 | 1572 | 76 | |||
| XM_015114924.2 | XP_014970410.2 | 575 | 76 | |||
| XM_015114925.2 | XP_014970411.2 | 1599 | 76 | |||
| XM_028833709.1 | XP_028689542.1 | 2510 | 76 | |||
| XM_028833710.1 | XP_028689543.1 | 1976 | 76 | |||
| GenBank mRNA | JV728008.1 | - | 887 | 76 | ||
Set Membership: |
|