Search
Search

SLC9A7| Gene Symbol: SLC9A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127281.2 | XP_014982767.2 | 1663 | 63 | ||
| XM_028842472.1 | XP_028698305.1 | 1662 | 63 | |||
| GenBank mRNA | JU495016.1 | - | 1373 | 63 | ||
| JV663681.1 | - | 41 | 63 | |||
Set Membership: |
|