Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh07213333_g1
See other SUPT20H GE Assays ›
Gene Symbol:
SUPT20H
Gene Name:
SPT20 homolog, SAGA complex component
Gene Aliases:
FAM48A
Chromosome Location:
Chr.17: 15846771 - 15895883 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh07213333_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SUPT20H
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015120926.2
XP_014976412.1
817
107
XM_015120927.2
XP_014976413.1
1156
107
XM_015120929.2
XP_014976415.1
684
107
XM_015120930.2
XP_014976416.1
814
107
XM_015120931.2
XP_014976417.1
1153
107
XM_015120932.2
XP_014976418.1
865
107
XM_015120933.2
XP_014976419.1
1183
107
XM_015120934.2
XP_014976420.1
817
107
XM_015120936.2
XP_014976422.1
781
107
XM_015120937.2
XP_014976423.1
832
107
XM_015120939.2
XP_014976425.1
778
107
XM_015120940.2
XP_014976426.1
829
107
XM_015120941.2
XP_014976427.1
817
107
XM_015120942.2
XP_014976428.1
817
107
XM_015120943.2
XP_014976429.1
814
107
XM_015120944.2
XP_014976430.1
817
107
XM_015120945.2
XP_014976431.1
817
107
XM_015120946.2
XP_014976432.1
814
107
XM_015120947.2
XP_014976433.1
817
107
XM_015120948.2
XP_014976434.1
817
107
XM_015120949.2
XP_014976435.1
817
107
XM_015120950.2
XP_014976436.1
1156
107
XM_015120951.1
XP_014976437.1
898
107
XM_015120952.2
XP_014976438.1
814
107
XM_015120953.2
XP_014976439.1
817
107
XM_028837428.1
XP_028693261.1
1086
107
XM_028837429.1
XP_028693262.1
1186
107
XM_028837430.1
XP_028693263.1
690
107
XM_028837431.1
XP_028693264.1
926
107
XM_028837433.1
XP_028693266.1
1050
107
XM_028837435.1
XP_028693268.1
1147
107
XM_028837436.1
XP_028693269.1
817
107
GenBank mRNA
JU335347.1
-
760
107
JU335348.1
-
760
107
JU476308.1
-
718
107
JU476309.1
-
718
107
JV045758.1
-
735
107
JV047582.1
-
781
107
JV047583.1
-
781
107
JV718351.1
-
787
107
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Macaca mulatta
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline