Search
Search

NHSL1| Gene Symbol: NHSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095017.4 | XP_001095017.4 | 1143 | 111 | ||
| XM_001095123.4 | XP_001095123.4 | 726 | 111 | |||
| XM_015137357.2 | XP_014992843.2 | 1150 | 111 | |||
| XM_015137359.2 | XP_014992845.2 | 909 | 111 | |||
| GenBank mRNA | JV066473.1 | - | 236 | 111 | ||
Set Membership: |
|