Search
Search

PLEKHG5| Gene Symbol: PLEKHG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141788.2 | XP_014997274.2 | 2021 | 64 | ||
| XM_028840525.1 | XP_028696358.1 | 2103 | 64 | |||
| XM_028840528.1 | XP_028696361.1 | 2103 | 64 | |||
| XM_028840532.1 | XP_028696365.1 | 2058 | 64 | |||
| XM_028840535.1 | XP_028696368.1 | 2513 | 64 | |||
| XM_028840538.1 | XP_028696371.1 | 2610 | 64 | |||
| XM_028840542.1 | XP_028696375.1 | 2066 | 64 | |||
| XM_028840545.1 | XP_028696378.1 | 2168 | 64 | |||
| XM_028840547.1 | XP_028696380.1 | 2676 | 64 | |||
| XM_028840550.1 | XP_028696383.1 | 2424 | 64 | |||
| XM_028840554.1 | XP_028696387.1 | 2513 | 64 | |||
| XM_028840559.1 | XP_028696392.1 | 2066 | 64 | |||
| XM_028840565.1 | XP_028696398.1 | 2103 | 64 | |||
| GenBank mRNA | JV588300.1 | - | 2001 | 64 | ||
| JV600305.1 | - | 1961 | 64 | |||
| JV690773.1 | - | 1999 | 64 | |||
Set Membership: |
|