Search
Search

YTHDC2| Gene Symbol: YTHDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849660.1 | XP_028705493.1 | 3046 | 102 | ||
| XM_028849661.1 | XP_028705494.1 | 2875 | 102 | |||
| XM_028849662.1 | XP_028705495.1 | 2675 | 102 | |||
| GenBank mRNA | JU327508.1 | - | 2459 | 102 | ||
| JU471279.1 | - | 2993 | 102 | |||
Set Membership: |
|