Search
Search

NSD3| Gene Symbol: NSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145177.2 | XP_015000663.2 | 4613 | 73 | ||
| XM_015145179.2 | XP_015000665.2 | 4613 | 73 | |||
| XM_015145180.2 | XP_015000666.2 | 4586 | 73 | |||
| XM_015145181.2 | XP_015000667.2 | 4586 | 73 | |||
| XM_015145182.2 | XP_015000668.2 | 4439 | 73 | |||
| GenBank mRNA | JU335145.1 | - | 3392 | 73 | ||
| JU476148.1 | - | 3609 | 73 | |||
| JV654715.1 | - | 268 | 73 | |||