Search
Search

DPT| Gene Symbol: DPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125060.2 | XP_014980546.1 | 477 | 74 | ||
| GenBank mRNA | JU474560.1 | - | 436 | 74 | ||
| JV043957.1 | - | 458 | 74 | |||
| JV635999.1 | - | 458 | 74 | |||
Set Membership: |
|