Search
Search

ACSL6| Gene Symbol: ACSL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140720.2 | XP_014996206.1 | 2124 | 92 | ||
| XM_015140721.2 | XP_014996207.1 | 2124 | 92 | |||
| XM_015140722.2 | XP_014996208.1 | 2158 | 92 | |||
| XM_015140723.2 | XP_014996209.1 | 2255 | 92 | |||
| XM_015140726.2 | XP_014996212.1 | 2034 | 92 | |||
| XM_028849707.1 | XP_028705540.1 | 2046 | 92 | |||
| XM_028849708.1 | XP_028705541.1 | 2202 | 92 | |||
| GenBank mRNA | JU321991.1 | - | 2083 | 92 | ||
| JU321992.1 | - | 2083 | 92 | |||
| JV046560.1 | - | 2050 | 92 | |||
| JV452516.1 | - | 2060 | 92 | |||
| JV452517.1 | - | 2060 | 92 | |||
Set Membership: |
|