Search
Search

KIAA1841| Gene Symbol: KIAA1841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799259.3 | XP_002799305.2 | 488 | 135 | ||
| XM_028832458.1 | XP_028688291.1 | 488 | 135 | |||
| XM_028832459.1 | XP_028688292.1 | 488 | 135 | |||
| GenBank mRNA | JU322642.1 | - | 381 | 135 | ||
| JU494930.1 | - | 427 | 135 | |||
Set Membership: |
|