Search
Search

TSTD2| Gene Symbol: TSTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117235.2 | XP_014972721.2 | 991 | 110 | ||
| GenBank mRNA | JU323303.1 | - | 908 | 110 | ||
| JU323304.1 | - | 908 | 110 | |||
| JV636564.1 | - | 916 | 110 | |||
Set Membership: |
|