Search
Search

DAGLB| Gene Symbol: DAGLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133252.2 | XP_014988738.2 | 566 | 65 | ||
| GenBank mRNA | JU333415.1 | - | 443 | 65 | ||
| JV609015.1 | - | 507 | 65 | |||
Set Membership: |
|