Search
Search

PRRC2B| Gene Symbol: PRRC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834709.1 | XP_028690542.1 | 2045 | 52 | ||
| XM_028834710.1 | XP_028690543.1 | 2045 | 52 | |||
| XM_028834711.1 | XP_028690544.1 | 2042 | 52 | |||
| XM_028834712.1 | XP_028690545.1 | 2039 | 52 | |||
| XM_028834713.1 | XP_028690546.1 | 2044 | 52 | |||
| GenBank mRNA | JU336768.1 | - | 1971 | 52 | ||
| JU477528.1 | - | 1971 | 52 | |||
| JV048472.1 | - | 1970 | 52 | |||
| JV048473.1 | - | 1964 | 52 | |||
| JV565196.1 | - | 1971 | 52 | |||
Set Membership: |
|