Search
Search

BRPF3| Gene Symbol: BRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136245.2 | XP_014991731.1 | 2238 | 108 | ||
| XM_015136246.2 | XP_014991732.1 | 1899 | 108 | |||
| XM_028847624.1 | XP_028703457.1 | 1996 | 108 | |||
| XM_028847626.1 | XP_028703459.1 | 2405 | 108 | |||
| GenBank mRNA | JU334003.1 | - | 1891 | 108 | ||
| JU475188.1 | - | 1903 | 108 | |||
| JV046769.1 | - | 1987 | 108 | |||
| JV712156.1 | - | 1862 | 108 | |||
Set Membership: |
|