Search
Search

METTL16| Gene Symbol: METTL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835870.1 | XP_028691703.1 | 488 | 101 | ||
| XM_028835872.1 | XP_028691705.1 | 335 | 101 | |||
| GenBank mRNA | JU493269.1 | - | 452 | 101 | ||
| JV661183.1 | - | 454 | 101 | |||
Set Membership: |
|