Search
Search

DPYSL4| Gene Symbol: DPYSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148475.2 | XP_015003961.1 | 1261 | 84 | ||
| GenBank mRNA | JU332538.1 | - | 1140 | 84 | ||
| JU472353.1 | - | 1230 | 84 | |||
| JV045942.1 | - | 1162 | 84 | |||
| JV602005.1 | - | 1242 | 84 | |||
Set Membership: |
|