Search
Search

SETX| Gene Symbol: SETX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116503.2 | XP_014971989.2 | 7101 | 97 | ||
| XM_015116511.2 | XP_014971997.2 | 7101 | 97 | |||
| XM_028834686.1 | XP_028690519.1 | 7101 | 97 | |||
| XM_028834687.1 | XP_028690520.1 | 7157 | 97 | |||
| XM_028834689.1 | XP_028690522.1 | 7040 | 97 | |||
| XM_028834690.1 | XP_028690523.1 | 7041 | 97 | |||
| XM_028834691.1 | XP_028690524.1 | 6991 | 97 | |||
| XM_028834692.1 | XP_028690525.1 | 7167 | 97 | |||
| XM_028834693.1 | XP_028690526.1 | 7101 | 97 | |||
| XM_028834694.1 | XP_028690527.1 | 7167 | 97 | |||
| XM_028834695.1 | XP_028690528.1 | 7101 | 97 | |||
| XM_028834696.1 | XP_028690529.1 | 7101 | 97 | |||
| XM_028834698.1 | XP_028690531.1 | 6208 | 97 | |||
| GenBank mRNA | JU333536.1 | - | 7026 | 97 | ||
| JU474802.1 | - | 7022 | 97 | |||
Set Membership: |
|