Search
Search

TAF1C| Gene Symbol: TAF1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126706.2 | XP_014982192.2 | 661 | 121 | ||
| XM_015126707.2 | XP_014982193.2 | 661 | 121 | |||
| GenBank mRNA | JU336987.1 | - | 611 | 121 | ||
| JU477712.1 | - | 634 | 121 | |||
| JV726318.1 | - | 625 | 121 | |||
Set Membership: |
|