Search
Search

MTREX| Gene Symbol: MTREX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140078.2 | XP_014995564.1 | 1461 | 80 | ||
| GenBank mRNA | JU322316.1 | - | 1407 | 80 | ||
| JU322322.1 | - | 1407 | 80 | |||
| JU471208.1 | - | 1295 | 80 | |||
| JV043736.1 | - | 1474 | 80 | |||
| JV046600.1 | - | 1474 | 80 | |||
| JV710895.1 | - | 1486 | 80 | |||
Set Membership: |
|