Search
Search

BTAF1| Gene Symbol: BTAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147682.2 | XP_015003168.2 | 552 | 110 | ||
| XM_015147683.2 | XP_015003169.2 | 548 | 110 | |||
| XM_015147684.2 | XP_015003170.2 | 548 | 110 | |||
| XM_015147685.2 | XP_015003171.1 | 547 | 110 | |||
| XM_028826656.1 | XP_028682489.1 | 627 | 110 | |||
| GenBank mRNA | JU337000.1 | - | 337 | 110 | ||
| JU477727.1 | - | 321 | 110 | |||
| JU477728.1 | - | 321 | 110 | |||
| JV562308.1 | - | 343 | 110 | |||
Set Membership: |
|