Search
Search

PPA1| Gene Symbol: PPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147456.2 | XP_015002942.1 | 229 | 125 | ||
| GenBank mRNA | JU320391.1 | - | 87 | 125 | ||
| JU473409.1 | - | 87 | 125 | |||
| JV044658.1 | - | 86 | 125 | |||
| JV638679.1 | - | 87 | 125 | |||
Set Membership: |
|