Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh07217647_m1
See other DEPDC5 GE Assays ›
Gene Symbol:
DEPDC5
Gene Name:
DEP domain containing 5, GATOR1 subcomplex subunit
Gene Aliases:
EGK_02963
Chromosome Location:
Chr.10: 19594025 - 19758791 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh07217647_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DEPDC5
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015150093.2
XP_015005579.2
3003
117
XM_015150094.2
XP_015005580.2
2907
117
XM_015150096.2
XP_015005582.2
3003
117
XM_015150099.2
XP_015005585.2
2769
117
XM_015150100.2
XP_015005586.2
2744
117
XM_015150101.2
XP_015005587.2
2673
117
XM_028827770.1
XP_028683603.1
3029
117
XM_028827771.1
XP_028683604.1
2931
117
XM_028827772.1
XP_028683605.1
3029
117
XM_028827773.1
XP_028683606.1
3029
117
XM_028827774.1
XP_028683607.1
3002
117
XM_028827775.1
XP_028683608.1
2880
117
XM_028827776.1
XP_028683609.1
3029
117
XM_028827777.1
XP_028683610.1
2907
117
XM_028827778.1
XP_028683611.1
3002
117
XM_028827779.1
XP_028683612.1
2880
117
XM_028827780.1
XP_028683613.1
2795
117
XM_028827781.1
XP_028683614.1
2673
117
XM_028827782.1
XP_028683615.1
2768
117
XM_028827783.1
XP_028683616.1
2684
117
XM_028827784.1
XP_028683617.1
2450
117
GenBank mRNA
JU337272.1
-
2834
117
JV048812.1
-
2916
117
JV637100.1
-
2902
117
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Macaca mulatta
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline