Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh07217655_m1
See other DEPDC5 GE Assays ›
Gene Symbol:
DEPDC5
Gene Name:
DEP domain containing 5, GATOR1 subcomplex subunit
Gene Aliases:
EGK_02963
Chromosome Location:
Chr.10: 19594025 - 19758791 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh07217655_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DEPDC5
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015150093.2
XP_015005579.2
4415
86
XM_015150094.2
XP_015005580.2
4319
86
XM_015150096.2
XP_015005582.2
4349
86
XM_015150099.2
XP_015005585.2
4181
86
XM_015150100.2
XP_015005586.2
4156
86
XM_015150101.2
XP_015005587.2
4019
86
XM_028827770.1
XP_028683603.1
4441
86
XM_028827771.1
XP_028683604.1
4343
86
XM_028827772.1
XP_028683605.1
4441
86
XM_028827773.1
XP_028683606.1
4441
86
XM_028827774.1
XP_028683607.1
4414
86
XM_028827775.1
XP_028683608.1
4292
86
XM_028827776.1
XP_028683609.1
4375
86
XM_028827777.1
XP_028683610.1
4253
86
XM_028827778.1
XP_028683611.1
4348
86
XM_028827779.1
XP_028683612.1
4226
86
XM_028827780.1
XP_028683613.1
4207
86
XM_028827781.1
XP_028683614.1
4085
86
XM_028827782.1
XP_028683615.1
4180
86
XM_028827783.1
XP_028683616.1
4096
86
XM_028827784.1
XP_028683617.1
3862
86
GenBank mRNA
JU337272.1
-
4180
86
JV048812.1
-
4262
86
JV637100.1
-
4248
86
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Macaca mulatta
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline