Search
Search

JAKMIP3| Gene Symbol: JAKMIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148474.2 | XP_015003960.2 | 1376 | 99 | ||
| XM_028827023.1 | XP_028682856.1 | 1385 | 99 | |||
| XM_028827024.1 | XP_028682857.1 | 1389 | 99 | |||
| XM_028827025.1 | XP_028682858.1 | 1383 | 99 | |||
| XM_028827026.1 | XP_028682859.1 | 1383 | 99 | |||
| XM_028827027.1 | XP_028682860.1 | 1376 | 99 | |||
| XM_028827028.1 | XP_028682861.1 | 1160 | 99 | |||
| XM_028827029.1 | XP_028682862.1 | 1153 | 99 | |||
| XM_028827030.1 | XP_028682863.1 | 1157 | 99 | |||
| GenBank mRNA | JU331071.1 | - | 964 | 99 | ||
| JU342026.1 | - | 964 | 99 | |||