Search
Search

TBCD| Gene Symbol: TBCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837041.1 | XP_028692874.1 | 1668 | 99 | ||
| XM_028837042.1 | XP_028692875.1 | 1668 | 99 | |||
| GenBank mRNA | JU336114.1 | - | 1651 | 99 | ||
| JU470766.1 | - | 1672 | 99 | |||
| JV044894.1 | - | 1639 | 99 | |||
Set Membership: |
|