Search
Search

WDR62| Gene Symbol: WDR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123728.2 | XP_014979214.1 | 4267 | 82 | ||
| XM_015123729.2 | XP_014979215.1 | 4252 | 82 | |||
| XM_015123730.2 | XP_014979216.1 | 4198 | 82 | |||
| XM_028839605.1 | XP_028695438.1 | 4156 | 82 | |||
| XM_028839606.1 | XP_028695439.1 | 4104 | 82 | |||
| GenBank mRNA | JU472486.1 | - | 4231 | 82 | ||
Set Membership: |
|